Invitrogen
Manufacturer Code:PA579435
Catalog # PIPA579435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human IDE recombit protein (Position: F485-K756). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abeta-degrading protease; Insulin protease; Insulin-degrading enzyme; Insulin-degrading enzyme Abeta-degrading protease Insulin protease Insulinase Insulysin IDE; Insulinase; Insulysin
Gene Aliases: 1300012G03Rik; 4833415K22Rik; AA675336; AI507533; IDE; INSDEGM; INSULYSIN
UniProt ID: (Human) P14735, (Rat) P35559
Entrez Gene ID: (Human) 3416, (Rat) 25700, (Mouse) 15925
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.