Invitrogen
Manufacturer Code:PA511520
Catalog # PIPA511520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 406-435 amino acids from the central region of human IDE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abeta-degrading protease; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin; Insulysin Insulinase Insulin protease
Gene Aliases: IDE; INSULYSIN
UniProt ID: (Human) P14735
Entrez Gene ID: (Human) 3416
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.