OriGene
Manufacturer Code:TA700103
Catalog # 50-167-7639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D1 |
Immunogen | Full length human recombit protein of human ID3 produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHb25; Class B basic helix-loop-helix protein 25; DNA-binding protein inhibitor ID-3; HEIR-1 bHLHb25; Helix-loop-helix protein HEIR-1; ID-like protein inhibitor HLH 1R21; Inhibitor of differentiation 3; Inhibitor of DNA binding 3; inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Gene Aliases: 1R21; BHLHB25; HEIR-1; HEIR1; ID3
UniProt ID: (Human) Q02535
Entrez Gene ID: (Human) 3399
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.