Invitrogen
Manufacturer Code:PA547617
Catalog # PIPA547617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse ID1 Met1-Glu135 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb24; BHLHB24 bHLHb24DNA-binding protein inhibitor ID-1 Class B basic helix-loop-helix protein 24 dJ857M17.1.2 dJ857M17.1.2 (inhibitor of DNA binding 1 domit negative helix-loop-helixprotein) ID inhibitor of differentiation 1 Inhibitor of DNA binding 1 inhibitor of DNA binding 1 domit negative helix-loop-helix protein; Class B basic helix-loop-helix protein 24; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; Inhibitor of differentiation 1; Inhibitor of DNA binding 1; inhibitor of DNA binding 1, dominant negative helix-loop-helix protein
Gene Aliases: AI323524; BHLHB24; D2Wsu140e; ID; ID1; Idb1
UniProt ID: (Human) P41134
Entrez Gene ID: (Human) 3397, (Mouse) 15901
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.