Invitrogen
Manufacturer Code:PA5111470
Catalog # PIPA5111470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ID1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323524 BHLHB24 Class B basic helix-loop-helix protein 24 D2Wsu140e dJ857M17.1.2 DNA-binding protein inhibitor ID-1 ID Id1 Id-1 ID125A Idb1 Inhibitor of differentiation 1 Inhibitor of DNA binding 1; bHLHb24; Class B basic helix-loop-helix protein 24; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; Inhibitor of differentiation 1; Inhibitor of DNA binding 1; inhibitor of DNA binding 1, dominant negative helix-loop-helix protein
Gene Aliases: BHLHB24; ID; ID1
UniProt ID: (Human) P41134
Entrez Gene ID: (Human) 3397
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.