Invitrogen
Manufacturer Code:PA5100067
Catalog # PIPA5100067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ICT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110001A02Rik 1110002E03Rik 39S ribosomal protein L58 mitochondrial Digestion substraction 1 DS1 DS-1 Ict1 immature colon carcinoma transcript 1 immature colon carcinoma transcript 1 protein Immature colon carcinoma transcript 1 protein homolog Mitochondrial large ribosomal subunit protein mL62 mitochondrial ribosomal protein L58 Mrpl58 MRP-L58 peptidyl-tRNA hydrolase ICT1 mitochondrial; 39S ribosomal protein L58, mitochondrial; Digestion substraction 1; DS-1; Immature colon carcinoma transcript 1 protein; Immature colon carcinoma transcript 1 protein homolog; Mitochondrial large ribosomal subunit protein mL62; MRP-L58; Peptidyl-tRNA hydrolase ICT1, mitochondrial
Gene Aliases: 1110001A02Rik; 1110002E03Rik; DS-1; DS1; ICT1; MRP-L58; MRPL58
UniProt ID: (Mouse) Q8R035, (Human) B2RAD1
Entrez Gene ID: (Mouse) 68572, (Rat) 303673, (Human) 3396
Molecular Function: RNA binding protein nucleic acid binding translation factor translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.