Novus Biologicals
Manufacturer Code:NBP242633PECY7
Catalog # N242633PEC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1000000000 |
Immunogen | Recombinant human ICT1 (30-206aa) purified from E. coli |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: 39S ribosomal protein L58, mitochondrial; Digestion substraction 1; Digestion substraction 1 DS-1DS1 EC 3.1.1.29 immature colon carcinoma transcript 1 Immature colon carcinoma transcript 1 protein peptidyl-tRNA hydrolase ICT1 mitochondrial; DS-1; Immature colon carcinoma transcript 1 protein; Mitochondrial large ribosomal subunit protein mL62; MRP-L58; Peptidyl-tRNA hydrolase ICT1, mitochondrial
Gene Aliases: DS-1; DS1; ICT1; MRP-L58; MRPL58
UniProt ID: (Human) B2RAD1
Entrez Gene ID: (Human) 3396
Molecular Function: RNA binding protein nucleic acid binding translation factor translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.