Novus Biologicals
Manufacturer Code:NBP297509AF750
Catalog # NB102410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ICMT fragment. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.100 HSTE14 Isoprenylcysteine Carboxyl Methyltransferase Isoprenylcysteine carboxylmethyltransferase MST098 MSTP098 PCCMT PCMT PPMT Prenylated Protein Carboxyl Methyltransferase Protein-S-Isoprenylcysteine O-Methyltransferase; Isoprenylcysteine carboxylmethyltransferase; pcCMT; PPMT; Prenylated protein carboxyl methyltransferase; Prenylcysteine carboxyl methyltransferase; protein-S isoprenylcysteine O-methyltransferase; Protein-S-isoprenylcysteine O-methyltransferase
Gene Aliases: HSTE14; ICMT; MST098; MSTP098; PCCMT; PCMT; PPMT
UniProt ID: (Human) O60725
Entrez Gene ID: (Human) 23463
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.