Abnova
Manufacturer Code:H00023463K
Catalog # 89-200-609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ICMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ICMT peptide using ARM Technology.
For Research Use Only
Protein Aliases: HSTE14 MGC39955 MST098 MSTP098 PCCMT PCMT PPMT; Isoprenylcysteine carboxylmethyltransferase; pcCMT; PPMT; Prenylated protein carboxyl methyltransferase; Prenylcysteine carboxyl methyltransferase; protein-S isoprenylcysteine O-methyltransferase; Protein-S-isoprenylcysteine O-methyltransferase
Gene Aliases: HSTE14; ICMT; MST098; MSTP098; PCCMT; PCMT; PPMT
UniProt ID: (Human) O60725
Entrez Gene ID: (Human) 23463
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.