Invitrogen
Manufacturer Code:PA551513
Catalog # PIPA551513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ICK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ciliogenesis associated kinase 1; ECO LCK2 MRK MAK-related kinase hICK laryngeal cancer kinase 2 serine/threonine protein kinase serine/threonine-protein kinase ICK; hICK; Intestinal cell kinase; Laryngeal cancer kinase 2; LCK2; MAK-related kinase; MRK; serine/threonine protein kinase; Serine/threonine-protein kinase ICK
Gene Aliases: CILK1; ECO; ICK; KIAA0936; LCK2; MRK
UniProt ID: (Human) Q9UPZ9
Entrez Gene ID: (Human) 22858
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.