R&D Systems
Manufacturer Code:MAB3016SP
Catalog # MAB3016SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 536017 |
Immunogen | <em>E. coli</em>-derived recombinant human ICAT Met1-Gln81 Accession No. Q9NSA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: Beta-catenin-interacting protein 1; beta-catenin-interacting protein ICAT; beta-catenin-interacting protein ICAT catenin beta interacting protein 1 CTNNBIP1 ICATbeta-interacting protein 1 Inhibitor of beta-catenin and Tcf-4; catenin, beta interacting protein 1; Inhibitor of beta-catenin and Tcf-4
Gene Aliases: CTNNBIP1; ICAT
UniProt ID: (Human) Q9NSA3
Entrez Gene ID: (Human) 56998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.