Invitrogen
Manufacturer Code:PA542734
Catalog # PIPA542734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ICAM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ICAM-5; intercellular adhesion molecule 3; Intercellular adhesion molecule 5; Intercellular adhesion molecule 5 telencephalin TLCN TLN; intercellular adhesion molecule 5, telencephalin; Telencephalin
Gene Aliases: CD50; ICAM-3; Icam3; ICAM5; TLCN; TLN
UniProt ID: (Human) Q9UMF0, (Rabbit) Q28730, (Mouse) Q60625
Entrez Gene ID: (Human) 7087, (Bovine) 505158, (Dog) 484958, (Horse) 100146897, (Rabbit) 100328960, (Mouse) 15898, (Rat) 313785
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.