R&D Systems
Manufacturer Code:FAB8397N025
Catalog # FAB8397N025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 729632 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4 Ala23-Ala240 Accession # Q14773 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD242; CD242 antigen; CD242 CD242 antigen ICAM4 ICAM-4 intercellular adhesion molecule 4 intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) intercellular adhesion molecule 4 (LW blood group) intercellular adhesion molecule 4 Landsteiner-Wiener blood group Landsteiner-Wiener blood group Landsteiner-Wiener blood group antigen a Landsteiner-Wiener blood group glycoprotein LWLW blood group protein; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW blood group protein
Gene Aliases: CD242; ICAM4; LW
UniProt ID: (Human) Q14773
Entrez Gene ID: (Human) 3386
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.