Invitrogen
Manufacturer Code:PA577215
Catalog # PIPA577215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ICAM-4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD242; CD242 antigen; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (LW blood group); Intercellular adhesion molecule 4 ICAM-4 Landsteiner-Wiener blood group glycoprotein LW blood group protein CD242 ICAM4 LW; intercellular adhesion molecule 4 isoform 3 anitgen LWa; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW blood group protein; LW glycoprotein
Gene Aliases: 1810015M19Rik; CD242; ICAM-4; ICAM4; LW
UniProt ID: (Human) Q14773, (Mouse) Q9ERM2
Entrez Gene ID: (Human) 3386, (Mouse) 78369, (Rat) 298702
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.