Invitrogen
Manufacturer Code:PA537869
Catalog # PIPA537869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RHGSRVIYSESLER from the internal region of ICAM4 (aa169-182) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD242; CD242 antigen; ICAM-4; ICAM4 intercellular adhesion molecule 4 Landsteiner-Wiener blood group LW CD242 antigen LW blood group protein Landsteiner-Wiener blood group antigen a Landsteiner-Wiener blood group glycoprotein intercellular adhesion molecule 4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW blood group protein
Gene Aliases: CD242; ICAM4; LW
UniProt ID: (Human) Q14773
Entrez Gene ID: (Human) 3386
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.