Invitrogen
Manufacturer Code:PA579430
Catalog # PIPA579430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse ICAM1 recombit protein (Position: G198-P537). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD54; ICAM-1; Intercellular adhesion molecule 1; Intercellular adhesion molecule 1 ICAM-1 MALA-2 MyD10 CD54 Icam1 Icam-1; MALA-2; MyD10
Gene Aliases: CD54; ICAM; Icam-1; Icam1; Ly-47; MALA-2
UniProt ID: (Mouse) P13597, (Rat) Q00238
Entrez Gene ID: (Mouse) 15894, (Rat) 25464
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.