Invitrogen
Manufacturer Code:MA529866
Catalog # PIMA529866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 37 |
Immunogen | Recombit Rat ICAM-1/CD54 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: BB2 P3.58 ICAM-1 cell surface glycoprotein P3.58 intercellular adhesion molecule 1 human rhinovirus receptor major group rhinovirus receptor; CD54; ICAM-1; Intercellular adhesion molecule 1
Gene Aliases: CD54; ICAM; Icam-1; Icam1
UniProt ID: (Rat) Q00238
Entrez Gene ID: (Rat) 25464
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.