Invitrogen
Manufacturer Code:MA180910
Catalog # PIMA180910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 15.2 |
Immunogen | Human monocytes. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BB2 P3.58 ICAM-1 cell surface glycoprotein P3.58 intercellular adhesion molecule 1 human rhinovirus receptor major group rhinovirus receptor; CD54; cell surface glycoprotein P3.58; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; Major group rhinovirus receptor
Gene Aliases: BB2; CD54; ICAM-1; ICAM1; P3.58
UniProt ID: (Human) P05362
Entrez Gene ID: (Pig) 396750, (Human) 3383
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.