Invitrogen
Manufacturer Code:MA524888
Catalog # PIMA524888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B10 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 1-331 of DFF45. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DFF-45; DFF45; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; DNA fragmentation factor, 45kDa, alpha polypeptide; H13 DFF-45 DFF1 ICAD DFF45 DNA fragmentation factor 45 kDa subunit DNA fragmentation factor subunit alpha inhibitor of CAD; ICAD; Inhibitor of CAD
Gene Aliases: DFF-45; DFF1; DFF45; DFFA; H13; ICAD
UniProt ID: (Human) O00273
Entrez Gene ID: (Human) 1676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.