Invitrogen
Manufacturer Code:PA129166
Catalog # PIPA129166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide - near the C terminus of mouse DFF45/ICAD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DFF-45; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; ICAD; Inhibitor of CAD; RP23-121D17.3 A330085O09Rik DFF35 Dff45 ICAD ICAD-L ICAD-S
Gene Aliases: A330085O09Rik; DFF35; Dff45; Dffa; ICAD; ICAD-L; ICAD-S
UniProt ID: (Mouse) O54786
Entrez Gene ID: (Mouse) 13347
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.