Invitrogen
Manufacturer Code:PA554895
Catalog # PIPA554895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human IBTK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bruton agammaglobulinemia tyrosine kinase inhibitor; BTK-binding protein; BTKI RP1-93K22.1 BTK-binding protein Bruton agammaglobulinemia tyrosine kinase inhibitor inhibitor of Bruton tyrosine kinase inhibitor of Bruton&primes tyrosine kinase inhibitor of Bruton&primes tyrosine kinase-alpha inhibitor of Bruton&primes tyrosine kinase-beta inhibitor of Brutonℙ IBtk; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma
Gene Aliases: BTBD26; BTKI; IBTK; KIAA1417
UniProt ID: (Human) Q9P2D0
Entrez Gene ID: (Human) 25998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.