Novus Biologicals
Manufacturer Code:NBP178061
Catalog # NBP178061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 1205-1217 of Human IBTK. IBTK is a currently uncharacterized human ankyrin repeat protein BAA92655. This protein has been suggested to bind to CUL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bruton agammaglobulinemia tyrosine kinase inhibitor; Bruton agammaglobulinemia tyrosine kinase inhibitor BTK-binding protein BTKI DKFZP564B116 IBtk inhibitor of Bruton agammaglobulinemia tyrosine kinase inhibitor of Bruton tyrosine kinase inhibitor of Bruton's tyrosine kinase inhibitor of Bruton's tyrosine kinase-alpha inhibitor of Bruton's tyrosine kinase-beta inhibitor of Bruton's tyrosine kinase-gamma KIAA1417 MGC142256 MGC142258 RP1-93K22.1; BTK-binding protein; IBtk; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma
Gene Aliases: BTBD26; BTKI; IBTK; KIAA1417
UniProt ID: (Human) Q9P2D0
Entrez Gene ID: (Human) 25998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.