Abnova
Manufacturer Code:H00025998K
Catalog # 89-207-251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human IBTK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human IBTK peptide using ARM Technology.
For Research Use Only
Protein Aliases: Bruton agammaglobulinemia tyrosine kinase inhibitor; BTK-binding protein; BTKI KIAA1417 MGC142256 MGC142258 RP1-93K22.1; IBtk; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma
Gene Aliases: BTBD26; BTKI; IBTK; KIAA1417
UniProt ID: (Human) Q9P2D0
Entrez Gene ID: (Human) 25998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.