Invitrogen
Manufacturer Code:MA538265
Catalog # PIMA538265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HL22-MS |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Iba1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI607846 AIF1 AIF-1 AIF-1 daintain Allograft inflammatory factor 1 allograft inflammatory factor-1 allograft inflammatory factor-1 splice variant G1 balloon angioplasty responsive transcript balloon angioplasty-responsive transcript 1 Bart1 BART-1 D17H6S50E DADB-70P7.8 daintain DASS-82G15.2 Em:AF129756.17 G1 Iba Iba1 interferon gamma responsive transcript ionized calcium binding adapter molecule 1 ionized calcium-binding adapter molecule 1 IRT1 IRT-1 Microglia response factor Mrf1 MRF-1 protein BART-1 protein G1 testis specific; AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; Ionized calcium-binding adapter molecule 1; Microglia response factor; MRF-1; protein BART-1; Protein G1
Gene Aliases: AIF-1; AIF1; BART-1; Bart1; G1; IBA1; IRT-1; IRT1; mrf-1; Mrf1
UniProt ID: (Human) P55008, (Rat) P55009
Entrez Gene ID: (Human) 199, (Rat) 29427
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.