Invitrogen
Manufacturer Code:MA529013
Catalog # PIMA529013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 145 |
Immunogen | Recombit Human IBA1/AIF1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AIF-1; Allograft inflammatory factor 1; Allograft inflammatory factor 1 Iba; interferon gamma responsive transcript; Ionized calcium-binding adapter molecule 1; Protein G1
Gene Aliases: AIF-1; AIF1; G1; IBA1; IRT-1; IRT1
UniProt ID: (Human) P55008
Entrez Gene ID: (Human) 199
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.