Invitrogen
Manufacturer Code:PA521274
Catalog # PIPA521274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 85 and 147 of Iba1 (Uniprot IDNo.P55008) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIF-1; Allograft inflammatory factor 1; DADB-70P7.8 AIF-1 IBA1 IRT-1 IRT1 allograft inflammatory factor-1 splice variant Hara-1 interferon gamma responsive transcript ionized calcium-binding adapter molecule 1 protein G1 Iba; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; Ionized calcium-binding adapter molecule 1; Protein G1; testis specific
Gene Aliases: AI607846; AIF-1; AIF1; D17H6S50E; G1; IBA1; IRT-1; IRT1
UniProt ID: (Human) P55008, (Mouse) O70200
Entrez Gene ID: (Human) 199, (Mouse) 11629
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.