Invitrogen
Manufacturer Code:PA5112150
Catalog # PIPA5112150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human IBA1/AIF1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI607846 AIF1 AIF-1 Allograft inflammatory factor 1 balloon angioplasty responsive transcript balloon angioplasty-responsive transcript 1 Bart1 BART-1 D17H6S50E DADB-70P7.8 daintain DASS-82G15.2 Em:AF129756.17 G1 Iba IBA1 interferon gamma responsive transcript ionized calcium binding adapter molecule 1 Ionized calcium-binding adapter molecule 1 IRT1 IRT-1 Microglia response factor Mrf1 MRF-1 protein BART-1 Protein G1 SBAB-548A10.7 testis specific; AIF-1; Allograft inflammatory factor 1; interferon gamma responsive transcript; Ionized calcium-binding adapter molecule 1; Protein G1
Gene Aliases: AIF-1; AIF1; G1; IBA1; IRT-1; IRT1
UniProt ID: (Human) P55008
Entrez Gene ID: (Human) 199
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.