Proteintech
Manufacturer Code:109041AP150UL
Catalog # 50-559-222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IBA1 Fusion Protein Ag1363 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIF 1 AIF1 G1 IBA1 IRT 1 Protein G1; AIF-1; Allograft inflammatory factor 1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; Ionized calcium-binding adapter molecule 1; Protein G1; testis specific
Gene Aliases: AI607846; AIF-1; AIF1; D17H6S50E; G1; IBA1; IRT-1; IRT1
UniProt ID: (Human) P55008, (Mouse) O70200
Entrez Gene ID: (Human) 199, (Mouse) 11629
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.