Novus Biologicals
Manufacturer Code:NBP259104
Catalog # NBP259104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Maps to a region between 962 and 1012 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.1.1 EC 6.1.1.5 FLJ10326 ILERS isoleucine tRNA ligase 2 mitochondrial Isoleucine--tRNA ligase isoleucine-tRNA synthetase 2 mitochondrial isoleucyl-tRNA synthetase 2 mitochondrial isoleucyl-tRNA synthetase mitochondrial mitochondrial isoleucine tRNA synthetase; IleRS; isoleucine tRNA ligase 2, mitochondrial; Isoleucine--tRNA ligase, mitochondrial; isoleucine-tRNA synthetase 2, mitochondrial; Isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase, mitochondrial; mitochondrial isoleucine tRNA synthetase
Gene Aliases: CAGSSS; IARS2; ILERS
UniProt ID: (Human) Q9NSE4
Entrez Gene ID: (Human) 55699
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.