Invitrogen
Manufacturer Code:PA578012
Catalog # PIPA578012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human IARS. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IRS; isoleucine tRNA ligase 1, cytoplasmic; isoleucine tRNA synthetase; Isoleucine--tRNA ligase, cytoplasmic; isoleucine-tRNA synthetase; isoleucyl tRNA synthetase; Isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase, cytoplasmic; PRO0785 IRS IARS1 ILERS isoleucyl-tRNA synthetase ILRS
Gene Aliases: IARS; IARS1; Iarsl; ILERS; ILRS; IRS; PRO0785
UniProt ID: (Human) P41252
Entrez Gene ID: (Human) 3376, (Rat) 306804
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.