Invitrogen
Manufacturer Code:PA544246
Catalog # PIPA544246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human IARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ileRS; IRS; isoleucine tRNA ligase 1, cytoplasmic; isoleucine tRNA synthetase; Isoleucine--tRNA ligase, cytoplasmic; isoleucine-tRNA synthetase; isoleucyl tRNA synthetase; Isoleucyl-tRNA synthetase; Isoleucyl-tRNA synthetase FLJ20736 IARS1 ILRS PRO0785 IRS ILERS; isoleucyl-tRNA synthetase, cytoplasmic
Gene Aliases: 2510016L12Rik; AI327140; AU044614; E430001P04Rik; fi46h05; IARS; IARS1; Iarsl; ILERS; ILRS; IRS; PRO0785; wu:fi46h05; zgc:63790
UniProt ID: (Human) P41252, (Mouse) Q8BU30
Entrez Gene ID: (Human) 3376, (Horse) 100054721, (Bovine) 509885, (Rabbit) 100356680, (Dog) 484216, (Pig) 100624746, (Rat) 306804, (Guinea pig) 100723914, (Mouse) 105148, (Zebrafish) 334393
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.