Novus Biologicals
Manufacturer Code:NBP159411
Catalog # NBP159411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PTPRN(protein tyrosine phosphatase receptor type N) The peptide sequence was selected from the middle region of PTPRN. Peptide sequence EVQQVPSPVSSEPPKAARPPVTPVLLEKKSPLGQSQPTVAGQPSARPAAE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA 512 EC 3.1.3.48 FLJ16131 IA2 IA-2 IA-2/PTP ICA 512 ICA3 ICA512 islet cell antigen 2 Islet cell antigen 512 Islet cell autoantigen 3 protein tyrosine phosphatase receptor type N protein tyrosine phosphatase-like N PTP IA-2 receptor-type tyrosine-protein phosphatase-like N R-PTP-N; ICA 512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; Islet cell antigen 512; Islet cell autoantigen 3; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase-like N; PTP IA-2; R-PTP-N; Receptor-type tyrosine-protein phosphatase-like N
Gene Aliases: IA-2; IA-2/PTP; IA2; ICA3; ICA512; PTPRN; R-PTP-N
UniProt ID: (Human) Q16849
Entrez Gene ID: (Human) 5798
Molecular Function: hydrolase phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.