Invitrogen
Manufacturer Code:PA520823
Catalog # PIPA520823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide from near the amino terminus of human IA-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IA-1 IA1 insulinoma-associated protein 1 zinc finger protein IA-1; insulinoma associated 1; insulinoma-associated 1; insulinoma-associated protein (IA-1); Insulinoma-associated protein 1; insulinoma-associated protein 1-like; Zinc finger protein IA-1
Gene Aliases: IA-1; IA1; INSM1
UniProt ID: (Human) Q01101, (Mouse) Q63ZV0
Entrez Gene ID: (Human) 3642, (Mouse) 53626, (Rat) 100912563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.