Novus Biologicals
Manufacturer Code:NBP290046B
Catalog # NB069283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Hydroxyacid Oxidase-1/HAO-1 (NP_060015.1 Leu 2-Ile 370). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: (S)-2-hydroxy-acid oxidase; (S)-2-hydroxy-acid oxidase EC 1.1.3.15 Glycolate oxidase GOX1MGC142227 GOXMGC142225 HAOX1 hydroxyacid oxidase (glycolate oxidase) 1 hydroxyacid oxidase 1; Glycolate oxidase; glycolate oxidase 1; GOX; HAOX1; hydroxyacid oxidase (glycolate oxidase) 1; Hydroxyacid oxidase 1
Gene Aliases: GOX; GOX1; HAO1; HAOX1
UniProt ID: (Human) Q9UJM8
Entrez Gene ID: (Human) 54363
Molecular Function: dehydrogenase isomerase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.