Novus Biologicals
Manufacturer Code:NBP132465
Catalog # NBP132465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human HYAL1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.1.35 FUS2 Hyal-1 hyaluronidase 1 hyaluronoglucosaminidase 1 Hyaluronoglucosaminidase-1 luCa-1 LUCA1hyaluronidase-1 Lung carcinoma protein 1 MGC45987 NAT6 plasma hyaluronidase tumor suppressor LUCA-1; Hyal-1; Hyaluronidase-1; Hyaluronoglucosaminidase-1; LuCa-1; Lung carcinoma protein 1; plasma hyaluronidase; tumor suppressor LUCA-1
Gene Aliases: HYAL-1; HYAL1; LUCA1; MPS9; NAT6
UniProt ID: (Human) Q12794
Entrez Gene ID: (Human) 3373
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.