Novus Biologicals
Manufacturer Code:NBP30381420UL
Catalog # NB094875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 80-390 of human Hyaluronan synthase 1 (NP_001514.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HA synthase 1; HA synthase 1 HAS huHAS1 hyaluronan synthase 1 hyaluronate synthase 1 hyaluronic acid synthase 1; Hyaluronan synthase 1; Hyaluronate synthase 1; Hyaluronic acid synthase 1
Gene Aliases: HAS; HAS1
UniProt ID: (Human) Q92839
Entrez Gene ID: (Human) 3036
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.