Novus Biologicals
Manufacturer Code:NBP202287
Catalog # NBP202287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI14F4 |
Immunogen | Full length human recombinant protein of human ELAVL1(NP_001410) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ELAV (embryonic lethal abnormal vision Drosophila)-like 1 (Hu antigen R) ELAV-like protein 1 embryonic lethal abnormal vision drosophila homolog-like 1 Hu antigen R Hua Hu-antigen R HuR HURELAV1 MelG; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV-like protein 1; embryonic lethal, abnormal vision, drosophila, homolog-like 1; Hu antigen R; Hu-antigen R; HuR
Gene Aliases: ELAV1; ELAVL1; Hua; HUR; MelG
UniProt ID: (Human) Q15717
Entrez Gene ID: (Human) 1994
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.