Proteintech
Manufacturer Code:130321AP150UL
Catalog # 50-172-8433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HuA/B/C/D Fusion Protein Ag3678 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like 1; ELAV like protein 4 ELAVL4 Hu antigen D Hu C/D HUD PNEM; ELAV like RNA binding protein 1; Elav-like generic protein; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 1; ELAV-like protein 2; ELAV-like protein 3; ELAV-like protein 4; elav-related A; elav-related C; elav-related D; elavC; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-like 2; embryonic lethal, abnormal vision-related A; embryonic lethal, abnormal vision-related C; embryonic lethal, abnormal vision-related D; etID309757.2; etID309927.3; Hu antigen C; Hu antigen D; Hu antigen R; HU-antigen A; HU-antigen B; HU-antigen C; HU-antigen D; Hu-antigen R; HuB; HuC; HuD; HuR; MelG; Nervous system-specific RNA-binding protein Hel-N1; Nervous system-specific RNA-binding protein Mel-N1; Paraneoplastic cerebellar degeneration-associated antigen; Paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; Protein elrD; RNA binding protein HuB; RNA-binding protein HUD3
Gene Aliases: 2410055N02Rik; 2600009P04Rik; Elav; ELAV1; Elav2; ELAVL1; ELAVL2; ELAVL3; ELAVL4; Elra; elrc; elrd; fa25f08; fb59a02; fc60g02; HEL-N1; HELN1; Hua; HUB; HUC; HUCL; HUD; HUR; id:ibd1248; mel-N1; MelG; mHuC; PLE21; PNEM; r-HuD; RGD1561943; W91709; wu:fa25f08; wu:fb59a02; wu:fb77b03; wu:fc24h01; wu:fc60g02; zgc:91918; zHuA; zHuC
UniProt ID: (Human) Q15717, (Human) Q12926, (Human) Q14576, (Human) P26378, (Rat) B5DF91, (Mouse) P70372, (Mouse) Q60899, (Mouse) Q60900, (Mouse) Q61701, (Rat) O09032, (Zebrafish) A0A0R4IEW8
Entrez Gene ID: (Human) 1994, (Human) 1993, (Human) 1995, (Human) 1996, (Rat) 363854, (Mouse) 15568, (Rat) 286973, (Mouse) 15569, (Rat) 282824, (Mouse) 15571, (Mouse) 15572, (Rat) 432358, (Zebrafish) 30736, (Zebrafish) 431719, (Zebrafish) 30732, (Zebrafish) 30737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.