Invitrogen
Manufacturer Code:PA569568
Catalog # PIPA569568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human Hs6st3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761K2315 heparan sulfate 6-O-sulfotransferase 3; Heparan-sulfate 6-O-sulfotransferase 3; heparan-sulfate 6-O-sulfotransferase 3 pseudogene; HS6ST-3
Gene Aliases: 6OST3; HS6ST-3; HS6ST3
UniProt ID: (Human) Q8IZP7, (Mouse) Q9QYK4
Entrez Gene ID: (Pig) 100620744, (Bovine) 538566, (Human) 266722, (Mouse) 50787
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.