Novus Biologicals
Manufacturer Code:NB11037253SS
Catalog # NB11037253T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). [Swiss-Prot P15304] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hormone-sensitive lipase; hormone-sensitive lipase hormone-sensitive lipase testicular isoform HSLEC 3.1.1.79 LHS lipase hormone-sensitive; hormone-sensitive lipase testicular isoform; HSL; lipase, hormone-sensitive; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: AOMS4; FPLD6; HSL; LHS; LIPE
UniProt ID: (Human) Q05469
Entrez Gene ID: (Human) 3991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.