R&D Systems
Manufacturer Code:AF7104SP
Catalog # AF7104SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Hormone-sensitive Lipase/HSL Met302-Leu425 Accession # Q05469 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Hormone-sensitive lipase; hormone-sensitive lipase Hormonesensitive Lipase hormone-sensitive lipase testicular isoform HSL HSLEC 3.1.1.79 LHS lipase hormone-sensitive LIPE; hormone-sensitive lipase testicular isoform; HSL; lipase, hormone-sensitive; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: AOMS4; FPLD6; HSL; LHS; LIPE
UniProt ID: (Human) Q05469
Entrez Gene ID: (Human) 3991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.