Invitrogen
Manufacturer Code:MA534735
Catalog # PIMA534735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG35-72 |
Immunogen | Recombinant protein to C-terminal human hnRNP Q. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610109K23Rik 4632417O19Rik Ab2-339 Glycine- and tyrosine-rich RNA-binding protein GRYRBP GRY-RBP Heterogeneous nuclear ribonucleoprotein Q hnRNP hnRNP Q HNRNPQ hnRNP-Q Hnrpq HNRPQ1 Liver regeneration-related protein LRRG077 NS1-associated protein 1 NSAP1 Nsap1l pp68 Synaptotagmin-binding cytoplasmic RNA-interacting protein SYNCRIP; Glycine- and tyrosine-rich RNA-binding protein; GRY-RBP; Heterogeneous nuclear ribonucleoprotein Q; hnRNP Q; NS1-associated protein 1; synaptotagmin binding, cytoplasmic RNA interacting protein; Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Gene Aliases: GRY-RBP; GRYRBP; hnRNP-Q; HNRNPQ; HNRPQ; HNRPQ1; NSAP1; PP68; SYNCRIP
UniProt ID: (Human) O60506
Entrez Gene ID: (Human) 10492
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.