Invitrogen
Manufacturer Code:MA535819
Catalog # PIMA535819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1656 |
Immunogen | A synthesized peptide derived from human hnRNP Q |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610109K23Rik 4632417O19Rik Ab2-339 Glycine- and tyrosine-rich RNA-binding protein GRYRBP GRY-RBP Heterogeneous nuclear ribonucleoprotein Q hnRNP hnRNP Q HNRNPQ hnRNP-Q Hnrpq HNRPQ1 Liver regeneration-related protein LRRG077 NS1-associated protein 1 NSAP1 Nsap1l pp68 Synaptotagmin-binding cytoplasmic RNA-interacting protein SYNCRIP; Glycine- and tyrosine-rich RNA-binding protein; GRY-RBP; Heterogeneous nuclear ribonucleoprotein Q; hnRNP Q; hnRNP-Q; Liver regeneration-related protein LRRG077; NS1-associated protein 1; pp68; RRM RNA binding protein GRY-RBP; synaptotagmin binding, cytoplasmic RNA interacting protein; Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Gene Aliases: 2610109K23Rik; 4632417O19Rik; Ab2-339; GRY-RBP; GRYRBP; hnRNP-Q; HNRNPQ; HNRPQ; HNRPQ1; NSAP1; Nsap1l; PP68; SYNCRIP
UniProt ID: (Human) O60506, (Rat) Q7TP47, (Mouse) Q7TMK9
Entrez Gene ID: (Human) 10492, (Rat) 363113, (Mouse) 56403
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.