Invitrogen
Manufacturer Code:PA5115532
Catalog # PIPA5115532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human macroH2A.1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Core histone macro-H2A.1; Core histone macro-H2A.1 H2A histone family member Y H2A histone family member Y H2A.y H2A/y H2AF12M H2AFY histone H2A.y Histone macroH2A1 histone macroH2A1.1 histone macroH2A1.2 MACROH2A1 MACROH2A1.1 macroH2A1.2 medulloblastoma antigen MU-MB-50.205 mH2A1; H2A histone family, member Y; H2A.y; H2A/y; Histone H2A.y; Histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; MACROH2A1.2; Medulloblastoma antigen MU-MB-50.205; mH2A1
Gene Aliases: H2A.y; H2A/y; H2AF12M; H2AFY; MACROH2A1; MACROH2A1.1; macroH2A1.2; mH2A1
UniProt ID: (Human) O75367, (Rat) Q02874, (Mouse) Q9QZQ8
Entrez Gene ID: (Human) 9555, (Rat) 29384, (Mouse) 26914
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.