Invitrogen
Manufacturer Code:PA517822
Catalog # PIPA517822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the amino-terminal sequence of human histone H4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG3379-PA; CG3379-PB; CG3379-PC; CG3379-PD; germinal histone H4; H4 histone family, member A; H4 histone family, member I; His4r-PA; His4r-PB; His4r-PC; His4r-PD; HIST1H4A H4/A H4FA Histone H4 Histone H4A; histone 1, H4a; histone 1, H4b; histone cluster 1, H4a; Histone H3; Histone H4; OGP; Osteogenic growth peptide; RH52884p
Gene Aliases: BcDNA:RH52884; BUR5; CG31611; CG3379; CG33869; CG33871; CG33873; CG33875; CG33877; CG33879; CG33881; CG33883; CG33885; CG33887; CG33889; CG33891; CG33893; CG33895; CG33897; CG33899; CG33901; CG33903; CG33905; CG33907; CG33909; Dmel\CG3379; Dmel_CG3379; FBtr0082962; H1ft; H4; H4-12; H4-16; H4-53; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4C3; H4C4; H4C5; H4C6; H4C8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4ft; H4r; HHT1; HHT2; His4; His4-88CD; His4:CG31611; His4:CG33869; His4:CG33871; His4:CG33873; His4:CG33875; His4:CG33877; His4:CG33879; His4:CG33881; His4:CG33883; His4:CG33885; His4:CG33887; His4:CG33889; His4:CG33891; His4:CG33893; His4:CG33895; His4:CG33897; His4:CG33899; His4:CG33901; His4:CG33903; His4:CG33905; His4:CG33907; His4:CG33909; His4R; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; HIST2H4; HIST2H4A; HIST2H4B; Hist4; HIST4H4; N2749; SIN2; YBR010W; YBR0201; YNL031C
UniProt ID: (Baker's yeast) P61830, (Human) Q6FGB8, (Human) P62805, (Fruit fly) P84040, (Mouse) P62806, (Rat) P62804
Entrez Gene ID: (Baker's yeast) 852295, (Human) 8359, (Human) 8366, (Fruit fly) 41773, (Mouse) 326619, (Rat) 679983, (Mouse) 326620, (Rat) 64627
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.