Novus Biologicals
Manufacturer Code:NB10056348SS
Catalog # NB10056348T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acid residues 15-30 of human Histone H4 GAKRHRKVLRDNIQGI. This sequence is conserved across mammalian invertebrate and plant species. An NCBI BLAST shows no cross-reactivity of this sequence with other known histones. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H4/A H4/B H4/C H4/D H4/E H4/G H4/H H4/I H4/J H4/K H4/M H4/N H4/O H4/p H4F2 H4FA H4FB H4FC H4FD H4FE H4FG H4FH H4FI H4FJ H4FK H4FM H4FN H4FO HIST1H4A HIST1H4B HIST1H4C HIST1H4D HIST1H4E HIST1H4F HIST1H4H HIST1H4I HIST1H4J HIST1H4K HIST1H4L HIST2H4 HIST2H4A HIST2H4B histone 4 H4 histone cluster 4 H4 histone H4 MGC24116; histone 4, H4; Histone H4
Gene Aliases: H4-16; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4C3; H4C4; H4C5; H4C6; H4C8; H4C9; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4
UniProt ID: (Human) A2VCL0
Entrez Gene ID: (Human) 121504
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.