Novus Biologicals
Manufacturer Code:NB211081SS
Catalog # NB211081SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Dot blot (DB) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This Histone H3 [ac Lys9 ac Lys14] antibody was raised against synthetic acetylated peptide surrounding Lysines 9 and 14 of human Histone H3.2 [Swiss Prot Q71DI3]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H3 H3 histone family 3A H3.3A H3.3B H3F2 H3F3 H3F3B H3FM H3K9ac HIST2H3C histone H3.3 MGC87782 MGC87783; H3 histone family, member M; H3 histone, family 2; H3-clustered histone 13; H3-clustered histone 14; H3-clustered histone 15; histone 2, H3c; Histone H3.2; Histone H3/m; Histone H3/o
Gene Aliases: H3; H3.2; H3/M; H3C13; H3C14; H3C15; H3F2; H3FM; H3FN; HIST2H3A; HIST2H3C; HIST2H3D
UniProt ID: (Human) Q71DI3
Entrez Gene ID: (Human) 126961
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.