Novus Biologicals
Manufacturer Code:NBP267685
Catalog # NBP267685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN07-30 |
Immunogen | This Histone H3 [Dimethyl Lys9] antibody (SN07-30) was raised against synthetic peptide surrounding K9 of Histone H3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: H3 H3 histone family 3A H3.3A H3.3B H3F2 H3F3 H3F3B H3FM H3K9Me2 HIST2H3C histone H3.3 MGC87782 MGC87783; H3 histone family, member T; H3/g; H3/t; histone 3, H3; Histone H3.1t; Histone H3.4
Gene Aliases: H3-4; H3.4; H3/g; H3FT; H3t; HIST3H3
UniProt ID: (Human) Q16695
Entrez Gene ID: (Human) 8290
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.