Invitrogen
Manufacturer Code:MA515150
Catalog # PIMA515150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E.960.2 |
Immunogen | Synthetic peptide corresponding to the carboxy-terminal residues of human histone H3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CG5825-PA; CG5825-PC; CG5825-PD; H3 histone family, member A; H3 histone, family 3A; H3.A; H3.B; H3L-like histone; His3.3A-PA; His3.3A-PC; His3.3A-PD; histone ; histone 1, H2ai; histone 1, H3a; histone cluster 1, H2ai; histone cluster 1, H2ai-like; histone cluster 1, H3a; histone cluster 3, H3; Histone H3 containing protein; Histone H3 Histone 1 H3a Histone H3.1 histone H3/a HIST1H3A; Histone H3.1; Histone H3.3; histone H3.3 ; Histone H3.3A; Histone H3.3B; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l; lamprey; lpy
Gene Aliases: CG5825; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; fb58e10; H3 histone; H3-3A; H3-3B; h3-5; H3.1-221; H3.1-291; H3.1-I; H3.3; H3.3A; H3.3B; H3/A; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3F3; H3F3A; H3F3B; h3f3b.1; h3f3c; h3f3d; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3g; H3h; H3i; His-3.3A; His3.3; His3.3A; His3.3B; Hist1h2ai; Hist1h2ail; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; I79_014844; PH3; PP781; wu:fb58e10; zgc:110292; zgc:56193; zgc:56418; zgc:64222; zgc:86731
UniProt ID: (Pig) Q71LE2, (Bovine) Q5E9F8, (Human) P84243, (Mouse) P84244, (Human) P68431, (Fruit fly) C0HL66, (Rat) Q6LED0, (Mouse) P68433, (Zebrafish) Q6PI20
Entrez Gene ID: (Pig) 396970, (Bovine) 326601, (Human) 3020, (Mouse) 15078, (Chinese hamster) 100753137, (Human) 8350, (Fruit fly) 33736, (Rat) 291159, (Rat) 679994, (Mouse) 360198, (Zebrafish) 406269
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.