Proteintech
Manufacturer Code:171681AP150UL
Catalog # 50-173-1414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Histone-H3 Fusion Protein Ag10644 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H3-clustered histone 13; H3-clustered histone 14; H3-clustered histone 15; H3/n H3/o H3F2 H3FM HIST2H3A HIST2H3C HIST2H3D histone cluster 2 H3a Histone H3 Histone H3.2 Histone H3/m Histone H3/o; histone 2, H3a; Histone H3.2; Histone H3/m; Histone H3/o
Gene Aliases: H3/n; H3/o; H3C13; H3C14; H3C15; H3F2; H3FM; HIST2H3A; HIST2H3C; HIST2H3D
UniProt ID: (Human) Q71DI3
Entrez Gene ID: (Human) 333932
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.